missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride, Mikromol

Product Code. 30461249
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461249 4 x 25 mg 100mg
30049404 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30461249 Supplier Mikromol Supplier No. MM0102.1000254AMP
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(Diethylamino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethyl-amino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride
CAS 857170-72-0
Analyte or Component Names 2-(Diethyl-amino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H22 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym Lidocaine Hydrochloride Imp. F (EP), Lidocaine Imp. F (EP), 2-(Diethyl-amino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. F (EP) as Hydrochloride, Lidocaine Hydrochloride Impurity F as Hydrochloride, Lidocaine Impurity F as Hydrochloride
InChI Formula InChI=1S/C14H22N2O.ClH/c1-5-16(6-2)10-14(17)15-13-9-7-8-11(3)12(13)4;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H
Solution Type Neat
SMILES Cl.CCN(CC)CC(=O)Nc1cccc(C)c1C
IUPAC Name 2-(diethylamino)-N-(2,3-dimethylphenyl)acetamide;hydrochloride
Molecular Weight (g/mol) 270.8
Quantity 4 x 25 mg
Formula Weight 270.1499 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.