missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate, TRC

Product Code. 30381666
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381666 100 mg 100mg
30385783 10 mg -
30385275 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381666 Supplier Toronto Research Chemicals Supplier No. TRCD680620100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-43-4
Molecular Formula C17H28N2O3
Molecular Weight (g/mol) 308.42
Synonym Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, 2-Diethylaminoethyl 4-amino-3-butoxybenzoate, 3-Butoxy-4-aminobenzoic acid 2-diethylaminoethyl ester, 4-Amino-3-butoxybenzoic acid 2-diethylaminoethyl ester, Benoxinate, Conjucain, Dorsacain, Novesinol, Oxybucaine, Oxybuprocaine, Prescaina
SMILES CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H28N2O3/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3/h8-9,13H,4-7,10-12,18H2,1-3H3
Formula Weight 308.21
Chemical Name or Material 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.