missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethanol, Mikromol

Product Code. 30464239
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30464239 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464239 Supplier Mikromol Supplier No. MM3771.040100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(Diethylamino)ethanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethylamino)ethanol
CAS 100-37-8
Analyte or Component Names 2-(Diethylamino)ethanol
Shipping Condition Room Temperature
Molecular Formula C6 H15 N O
Certifications/Compliance ISO 9001
Synonym 2-(Diethylamino)ethanol, (2-Hydroxyethyl)diethylamine, (Diethylamino)ethanol, 2-(Diethylamino)ethyl alcohol, 2-(N,N-Diethyl)ethanolamine, 2-(N,N-Diethylamino)ethanol, 2-Hydroxytriethylamine, A 22, A 22 (amine), Amino Alcohol 2A, DEAE, DEEA, Diethyl(2-hydroxyethyl)amine, Diethyl(β-hydroxyethyl)amine, Diethylethanolamine, Diethylmonoethanolamine, MKS, N,N-Diethyl(2-hydroxyethyl)amine, N,N-Diethyl-2-aminoethanol, N,N-Diethylethanolamine, N,N-Diethylmonoethanolamine, N-(2-Hydroxyethyl)diethylamine, NSC 8759, Pennad 150, β-(Diethylamino)ethanol
InChI Formula InChI=1S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3
Solution Type Neat
SMILES CCN(CC)CCO
IUPAC Name 2-(diethylamino)ethanol
Molecular Weight (g/mol) 117.19
Quantity 100 mg
Formula Weight 117.1154 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.