missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethanol, 99%, Thermo Scientific Chemicals

Product Code. 11447737
Change view
Click to view available options
Quantity:
1000 mL
5000 mL
Unit Size:
1000mL
5000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447737 1000 mL 1000mL
11457737 5000 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11447737 Supplier Thermo Scientific Alfa Aesar Supplier No. A16274.AU

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

Solubility
It is miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic , Avoid moisture. Keep away from strong oxidizing agents, strong acids, Copper, Zinc, Iron, Do not store near acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-37-8
Molecular Formula C6H15NO
MDL Number MFCD00002850
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
Synonym 2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
PubChem CID 7497
ChEBI CHEBI:52153
IUPAC Name 2-(diethylamino)ethanol
SMILES CCN(CC)CCO

Specifications

Melting Point -70°C
Density 0.883
pH 11.5
Boiling Point 161°C to 163°C
Flash Point 52°C (125°F)
Odor Amine-like
Linear Formula (CH3CH2)2NCH2CH2OH
Refractive Index 1.4415
Quantity 1000 mL
UN Number UN2686
Beilstein 741863
Sensitivity Air and light sensitive; Hygroscopic
Merck Index 14,3112
Solubility Information It is miscible in water.
Molecular Weight (g/mol) 117.192
Formula Weight 117.19
Percent Purity 99%
Chemical Name or Material 2-(Diethylamino)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.