missing translation for 'onlineSavingsMsg'
Learn More

2-Dibromomethyl-benzonitrile, TRC

Product Code. 30380610
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380610 250 mg 250mg
30374444 100 mg -
30383313 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380610 Supplier Toronto Research Chemicals Supplier No. TRCD425605250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 655-63-0
Molecular Formula C8H5Br2N
Molecular Weight (g/mol) 274.94
Synonym alpha,alpha-Dibromo-o-tolunitrile
SMILES BrC(Br)c1ccccc1C#N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H5Br2N/c9-8(10)7-4-2-1-3-6(7)5-11/h1-4,8H
Formula Weight 272.8789
Chemical Name or Material 2-Dibromomethyl-benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.