missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Di-tert-butylphosphino)ethylamine, 10% w/w soln. in THF

Product Code. 15435696
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435696 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435696 Supplier Alfa Aesar™ Supplier No. H60355.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1053658-84-6
Molecular Formula C10H24NP
Linear Formula [(CH3)3C]2PCH2CH2NH2
MDL Number MFCD11044862
Quantity 5g
UN Number UN2845
Synonym 2-di-t-butylphosphino ethylamine, 2-di-tert-butylphosphino ethylamine, 2-ditertbutylphosphino ethylamine, 2-aminoethyldi-tert-butylphosphine, 2-di-tert-butylphosphino ethanamine, 2-di-tert-butylphosphanyl ethan-1-amine, 2-aminoethyl di-tert-butyl phosphane, 2-di-tert-butylphosphino ethylamine, 10 wt. % in thf
InChI Key DZDGRVHIUYIMDS-UHFFFAOYSA-N
SMILES CC(C)(C)P(CCN)C(C)(C)C
IUPAC Name 2-ditert-butylphosphanylethanamine
Molecular Weight (g/mol) 189.283
PubChem CID 57553001
Formula Weight 189.28
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material 2-(Di-tert-butylphosphino)ethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.