missing translation for 'onlineSavingsMsg'
Learn More

2-[Di(methylthio)methylidene]malononitrile, TRC

Product Code. 30383790
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383790 500 mg
30371501 5 g
30388232 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383790 Supplier Toronto Research Chemicals Supplier No. TRCD462080500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5147-80-8
Molecular Formula C6H6N2S2
Molecular Weight (g/mol) 170.25
IUPAC Name 2-[bis(methylsulfanyl)methylidene]propanedinitrile
SMILES CSC(=C(C#N)C#N)SC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H6N2S2/c1-9-6(10-2)5(3-7)4-8/h1-2H3
Formula Weight 169.9972
Chemical Name or Material 2-[Di(methylthio)methylidene]malononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.