missing translation for 'onlineSavingsMsg'
Learn More

2-[Di(methylthio)methylidene]malononitrile, 97%, Thermo Scientific™

Product Code. 10190822
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190822 10 g Amber Glass Bottle 10g
10601103 1 g Amber Glass Bottle 1g
11306865 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190822 Supplier Thermo Scientific Maybridge Supplier No. CD00793EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5147-80-8
Molecular Formula C6H6N2S2
Molecular Weight (g/mol) 170.248
MDL Number MFCD00052730
InChI Key FICQFRCPSFCFBY-UHFFFAOYSA-N
Synonym 2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile
PubChem CID 99229
IUPAC Name 2-[bis(methylsulfanyl)methylidene]propanedinitrile
SMILES CSC(=C(C#N)C#N)SC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CN)2C=C(SCH3)2
Quantity 10 g
Formula Weight 170.26
Percent Purity 97%
Chemical Name or Material 2-[Di(methylthio)methylidene]malononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.