missing translation for 'onlineSavingsMsg'
Learn More

2-Desaminosulfonyl 3-Aminosulfonyl Dorzolamide, TRC

Product Code. 30357825
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357825 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357825 Supplier Toronto Research Chemicals Supplier No. TRCD28823510MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Desaminosulfonyl 3-Aminosulfonyl Dorzolamide
CAS 199734-71-9
Molecular Formula C10 H16 N2 O4 S3
Quantity 10 mg
Synonym (4S,6S)-4-(Ethylamino)-6-Methyl-5,6-Dihydro-4H-Thieno[2,3-B] Thiopyran-3-Sulfonamide 7,7-Dioxide
InChI Formula InChI=1S/C10H16N2O4S3/c1-3-12-7-4-6(2)18(13,14)10-9(7)8(5-17-10)19(11,15)16/h5-7,12H,3-4H2,1-2H3,(H2,11,15,16)/t6-,7-/m0/s1
SMILES CCN[C@H]1C[C@H](C)S(=O)(=O)c2scc(c12)S(=O)(=O)N
IUPAC Name (4S,6S)-4-(ethylamino)-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-3-sulfonamide
Molecular Weight (g/mol) 324.44
Formula Weight 324.0272
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.