missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxyuridine, TRC

Product Code. 30359767
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359767 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359767 Supplier Toronto Research Chemicals Supplier No. TRCD288010100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxyuridine
CAS 951-78-0
Molecular Formula C9H12N2O5
Quantity 100 mg
Synonym Uridine, 2'-deoxy- (6CI, 8CI, 9CI, ACI), 2'-Deoxyuridine (ACI), 1-(2-Deoxy-β-D-erythro-pentofuranosyl)uracil, 2,4(1H,3H)-Pyrimidinedione, 1-(2-deoxy-β-D-erythro-pentofuranosyl)-, 2,4(1H,3H)-Pyrimidinedione, 1-(2-deoxy-β-D-ribofuranosyl)-, 2'-Desoxyuridine, Deoxyribose uracil, Deoxyuridine, MeSH ID: D003857, NSC 23615, Uracil deoxyriboside, WV-NU-242A
InChI Formula InChI=1S/C9H12N2O5/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15/h1-2,5-6,8,12-13H,3-4H2,(H,10,14,15)/t5-,6+,8+/m0/s1
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N2C=CC(=O)NC2=O
Molecular Weight (g/mol) 228.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.