missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyuridine, 98+%

Product Code. p-7023007
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11478656

Brand: Thermo Scientific Chemicals A16026.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 951-78-0 | C9H12N2O5 | 228.20 g/mol

2'-Deoxyuridine is frequently halogenated to create thymidine analogues useful for studies of DNA synthesis and degradation mechanisms. Derivatized 2?-Deoxyuridines used as labeling substrates include chloro-2?-deoxyuridine (CldU), bromodeoxyuridine (BrdU) and/or iododeoxyuridine (IdU). Other useful analogues of 2?-deoxyuridine include 5-ethynyl-2?-deoxyuridine (DdU) and 5-hydroxymethyl-2?-deoxyuridine (HmdU). Laboratory suppression of deoxyuridine is used to diagnose megaloblastic anemias due to vitamin B12 and folate deficiencies. Deoxyuridine (dU) is used to indirectly determine if there are sufficient levels of folate and cobalamin in cell or tissue samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine is frequently halogenated to create thymidine analogues useful for studies of DNA synthesis and degradation mechanisms. Derivatized 2′-Deoxyuridines used as labeling substrates include chloro-2′-deoxyuridine (CldU), bromodeoxyuridine (BrdU) and/or iododeoxyuridine (IdU). Other useful analogues of 2′-deoxyuridine include 5-ethynyl-2′-deoxyuridine (DdU) and 5-hydroxymethyl-2′-deoxyuridine (HmdU). Laboratory suppression of deoxyuridine is used to diagnose megaloblastic anemias due to vitamin B12 and folate deficiencies. Deoxyuridine (dU) is used to indirectly determine if there are sufficient levels of folate and cobalamin in cell or tissue samples.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-78-0
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.20
MDL Number MFCD00006527
InChI Key MXHRCPNRJAMMIM-SHYZEUOFSA-N
Synonym 2'-deoxyuridine, deoxyuridine, uracil deoxyriboside, 2-deoxyuridine, 1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, deoxyribose uracil, uridine, 2'-deoxy, 2'-desoxyuridine, unii-w78i7ay22c, ccris 2832
PubChem CID 13712
ChEBI CHEBI:16450
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 164°C to 167°C
Quantity 1 g
Beilstein 24433
Sensitivity Air Sensitive
Merck Index 14,2910
Solubility Information Soluble in water.
Optical Rotation +52° (c=1.1 in 1N NaOH)
Formula Weight 228.21
Percent Purity 99%
Chemical Name or Material 2'-Deoxyuridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.