missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyuridine-5'-triphosphate Trisodium Salt >90% (contains ammonium salt), TRC

Product Code. 30352032
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
30352032 10 mg 10mg
30350082 100 mg 100mg
30358543 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30352032

Brand: Toronto Research Chemicals TRCD28803810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102814-08-4
Formula Weight 467.974
InChI Formula InChI=1S/C9H15N2O14P3.3Na/c12-5-3-8(11-2-1-7(13)10-9(11)14)23-6(5)4-22-27(18,19)25-28(20,21)24-26(15,16)17;;;/h1-2,5-6,8,12H,3-4H2,(H,18,19)(H,20,21)(H,10,13,14)(H2,15,16,17);;;/q;3*+1/p-3/t5-,6+,8+;;;/m0.../s1
IUPAC Name [[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate
Molecular Formula C9 H12 N2 O14 P3 . Na3
Molecular Weight (g/mol) 468.142
SMILES O=C(NC1=O)C=CN1[C@@H]2O[C@H](COP(OP(OP(O)(O[Na])=O)(O[Na])=O)(O[Na])=O)[C@@H](O)C2
Synonym 2'-deoxy-Uridine 5'-(tetrahydrogen triphosphate) trisodium salt, dUTP trisodium salt, dUTP sodium salt
Chemical Name or Material 2'-Deoxyuridine-5'-triphosphate Trisodium Salt

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.