missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxythymidine-5'-diphosphate trisodium salt, 97%

Product Code. 15470407
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470407 100 mg 100mg
15460407 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470407 Supplier Thermo Scientific Chemicals Supplier No. J64938.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95648-78-5 | C10H13N2Na3O11P2 | 468.13 g/mol

2'-Deoxythymidine-5'-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxythymidine-5′-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95648-78-5
Molecular Formula C10H13N2Na3O11P2
Molecular Weight (g/mol) 468.13
MDL Number MFCD00084748
InChI Key HBYPEEZBYHQLDM-SPSULGLQSA-K
PubChem CID 133109123
IUPAC Name trisodium;[[(2S,3R,5S)-3-hydroxy-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy-oxidophosphoryl] phosphate
SMILES [Na+].[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 468.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2'-Deoxythymidine-5'-diphosphate trisodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.