missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine, MedChemExpress

Product Code. 16489336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489336 Supplier Medchem Express Supplier No. HYW008638
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2’-deoxyadenosine inhibits the growth of human colon-carcinoma cell lines and is found to be associated with purine nucleoside phosphorylase (PNP) deficiency.

2’-deoxyadenosine inhibits the growth of human colon-carcinoma cell lines and is found to be associated with purine nucleoside phosphorylase (PNP) deficiency.

Specifications

Chemical Name or Material 2'-Deoxyinosine
CAS 890-38-0
Color White
Purity Grade Notes Research
Molecular Formula C10H12N4O4
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (198.23 mM; Need ultrasonic)
SMILES OC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)C3)C2=NC=N1
Molecular Weight (g/mol) 252.23
Formula Weight 252.23
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.