missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine 5'-Monophosphate Disodium Salt, TRC

Product Code. 30361271
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361271 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361271 Supplier Toronto Research Chemicals Supplier No. TRCD239805100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxyinosine 5'-Monophosphate Disodium Salt
CAS 14999-52-1
Molecular Formula C10H11N4Na2O7P
Quantity 100 mg
Synonym 2-Deoxyinosine-5-Monophosphate, Disodium Salt
InChI Formula InChI=1S/C10H13N4O7P.2Na/c15-5-1-7(21-6(5)2-20-22(17,18)19)14-4-13-8-9(14)11-3-12-10(8)16;;/h3-7,15H,1-2H2,(H,11,12,16)(H2,17,18,19);;/q;2*+1/p-2/t5-,6+,7+;;/m0../s1
SMILES [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP(=O)(O)[O-])n2cnc3c([O-])ncnc23
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(6-oxidopurin-9-yl)oxolan-2-yl]methyl hydrogen phosphate
Molecular Weight (g/mol) 376.17
Formula Weight 376.0161
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.