missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine, MedChemExpress

Product Code. 16468886
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468886 Supplier Medchem Express Supplier No. HYD0184

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2'-Deoxycytidine, a deoxyribonucleoside, could inhibit biological effects of Bromodeoxyuridine (Brdu).

2'-Deoxycytidine, a deoxyribonucleoside, could inhibit biological effects of Bromodeoxyuridine (Brdu).

Specifications

Chemical Name or Material 2'-Deoxycytidine
CAS 951-77-9
Color White
Purity Grade Notes Research
Molecular Formula C9H13N3O4
Quantity 100 mg
Synonym Deoxycytidine Cytosine deoxyriboside Deoxyribose cytidine
Solubility Information H2O : 50 mg/mL (220.05 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (220.05 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(N=C(C=C2)N)=O)O1)O
Molecular Weight (g/mol) 227.22
Formula Weight 227.22
Percent Purity 96.57%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.