missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine, 99+%, ACROS Organics™

£20.60 - £745.00

Chemical Identifiers

CAS 951-77-9
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-SHYZEUOFSA-N
Synonym 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
PubChem CID 13711
ChEBI CHEBI:15698
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)CO)O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10236380 SDS View Documents Acros Organics
226722500
250mg Glass bottle £20.60
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-01-2020
Please to purchase this item. Need a web account? Register with us today!
10104670 SDS View Documents Acros Organics
226725000
500mg Glass bottle £32.40
500mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-01-2020
Please to purchase this item. Need a web account? Register with us today!
10073060 SDS View Documents Acros Organics
226720010
1g Glass bottle £51.30
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-01-2020
Please to purchase this item. Need a web account? Register with us today!
10626334 SDS View Documents Acros Organics
226720050
5g Glass bottle £181.60
5g View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10076373 SDS View Documents Acros Organics
226720250
25g Glass bottle £745.00
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-01-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

951-77-9
227.22
CKTSBUTUHBMZGZ-SHYZEUOFSA-N
13711
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
C9H13N3O4
MFCD00006547
2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
CHEBI:15698
C1C(C(OC1N2C=CC(=NC2=O)N)CO)O

Specifications

2'-Deoxycytidine
Glass bottle
White
250mg
99.0
99% min. (HPLC)
MFCD00006547
Solubility in water: soluble
C1C(C(OC1N2C=CC(=NC2=O)N)CO)O
227.22
CHEBI:15698
Crystalline Powder
Authentic
8% max.
209°C to 211°C
951-77-9
100.0
C9H13N3O4
2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
CKTSBUTUHBMZGZ-SHYZEUOFSA-N
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
13711
227.22
99+%