missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine, 99+%

Product Code. 10104670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104670 500 mg Glass bottle 500mg
10236380 250 mg Glass bottle 250mg
10073060 1 g Glass bottle 1g
10626334 5 g Glass bottle 5g
10076373 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10104670 Supplier Thermo Scientific Chemicals Supplier No. 226725000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 951-77-9 | C9H13N3O4 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-77-9
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-JORGKRSHNA-N
Synonym 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
PubChem CID 13711
ChEBI CHEBI:15698
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 209°C to 211°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: soluble
Formula Weight 227.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.