missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxycytidine 5'-Monophosphate Hydrate, TRC

Product Code. 30359790
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359790 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359790 Supplier Toronto Research Chemicals Supplier No. TRCD2326752.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxycytidine 5'-Monophosphate Hydrate
CAS 1032-65-1
Molecular Formula C9H14N3O7P.xH2O
Quantity 2.5 g
Synonym 2'-Deoxy-5'-cytidylic Acid Hydrate, 2'-Deoxycytidine 5'-(Dihydrogen Phosphate) Hydrate
InChI Formula InChI=1S/C9H14N3O7P/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(19-8)4-18-20(15,16)17/h1-2,5-6,8,13H,3-4H2,(H2,10,11,14)(H2,15,16,17)/t5-,6+,8+/m0/s1
SMILES NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](COP(=O)(O)O)O2
Molecular Weight (g/mol) 307.2
Formula Weight 307.0569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.