missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine-5'-monophosphate, 98%

Product Code. 15476359
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15476359

Brand: Thermo Scientific Alfa Aesar J64402.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 653-63-4 | C10H14N5O6P | 331.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 1N ammonium hhydroxide (50 mg/ml) and water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-63-4
Molecular Formula C10H14N5O6P
Molecular Weight (g/mol) 331.23
MDL Number MFCD00005753
InChI Key KHWCHTKSEGGWEX-RRKCRQDMSA-N
Synonym damp, 2'-deoxyadenosine-5'-monophosphate, 2'-deoxyadenosine 5'-monophosphate, 2'-deoxyadenylic acid, deoxyadenylic acid, deoxy-amp, 2'-deoxy-amp, 2'-deoxyadenosine 5'-phosphate, 2'-damp, deoxyadenosine monophosphate
PubChem CID 12599
ChEBI CHEBI:17713
IUPAC Name [(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(O)=O)O3)C2=NC=N1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in 1N ammonium hhydroxide (50mg/ml) and water.
Formula Weight 331.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-monophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.