missing translation for 'onlineSavingsMsg'
Learn More

Melford 2-Deoxy-D-Ribose

Product Code. 30018233
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018233 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018233 Supplier Melford Supplier No. D25100100.0
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Deoxy-D-Ribose is a crucial sugar molecule in life sciences, particularly for its role in the structure of DNA.

2-Deoxy-D-Ribose is a crucial sugar molecule in life sciences, particularly for its role in the structure of DNA. Unlike ribose, which is found in RNA, 2-Deoxy-D-Ribose lacks an oxygen atom on the second carbon, giving DNA its name (deoxyribonucleic acid).This small structural difference provides DNA with greater stability compared to RNA.

Researchers study 2-Deoxy-D-Ribose to understand DNA synthesis, repair, and overall genetic regulation. Additionally, it is relevant in studying various biochemical pathways and disease mechanisms, including cancer and genetic disorders.

Specifications

CAS 533-67-5
Molecular Formula C5H10O4
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.