missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-glucose, MedChemExpress

Product Code. 16481389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481389 1 g 1g
16458567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481389 Supplier Medchem Express Supplier No. HY13966
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.

2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.

Specifications

Chemical Name or Material 2-Deoxy-D-glucose
CAS 154-17-6
Color White
Purity Grade Notes Research
Molecular Formula C6H12O5
Quantity 1 g
Synonym 2-DG 2-Deoxy-D-arabino-hexose D-Arabino-2-deoxyhexose
Solubility Information H2O : ≥ 24 mg/mL (146.20 mM)
SMILES OC1O[C@@H]([C@@H](O)[C@H](O)C1)CO
Molecular Weight (g/mol) 164.16
Formula Weight 164.16
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.