missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals

Product Code. p-4258923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10560371 1 g Glass bottle 1g
10152950 5 g Glass bottle 5g
11906901 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10560371 Supplier Thermo Scientific Chemicals Supplier No. 111980010
Add to basket
This item is not returnable. View return policy

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151328
InChI Key PMMURAAUARKVCB-PHUJZJCSNA-N
Synonym deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PubChem CID 17751002
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146.0°C to 147.0°C
Color White to Cream
Ignition Residue 1% max.
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,2905
Specific Rotation + 45.50
Solubility Information (2% in water) Clear colorless to light yellow
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Deoxy-D-glucose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.