missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 2-Deoxy-D-glucose
Description
2-Deoxy-D-glucose is a non-metabolizable glucose analog. Inhibits phosphorylation of glucose by hexokinase; causes depletion of cellular ATP. Also inhibits phosphoglucose isomerase (PGI) competitively. Causes cell cycle inhibition and cell death in in vitro models of hypoxia; blocks tumor cell growth in animal models. Also shown to induce the unfolded protein response (UPR). 2-Deoxy-D-glucose disrupts N-glycan expression on the surface of pancreatic adenocarcinoma cells. 2-Deoxy-D-glucose enhances chimeric antigen receptor T cell activity in different xenograft mouse models of pancreatic adenocarcinoma cells.
Specifications
Specifications
| CAS | 154-17-6 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C6H12O5 |
| Formula Weight | 164.16 |
| Purity | >98% |
| Molecular Weight (g/mol) | 164.16 |
| Chemical Name or Material | 2-Deoxy-D-glucose |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?