missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-Galactose, MP Biomedicals™

Product Code. 11442202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442202 Supplier MP Biomedicals Supplier No. 0215080001
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical reagent used as an inhibitor of fucosylation.

Inhibitor of fucosylation

Specifications

CAS 1949-89-9
Molecular Formula C6H12O5
MDL Number MFCD00014649
Quantity 1 g
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular Weight (g/mol) 164.16
PubChem CID 102191
ChEBI CHEBI:27411
Formula Weight 164.2
Grade Research
Physical Form Solid
Chemical Name or Material 2-Deoxy-D-Galactose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.