missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%

Product Code. 15406929
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406929 2 g 2g
15416929 5 g 5g
15426929 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406929 Supplier Thermo Scientific Chemicals Supplier No. J65482.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For studying galactose uptake into Escherichia coli | CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol

2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Melting Point 107°C to 110°C
Color White
Quantity 2 g
Beilstein 1723333
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Deoxy-D-galactose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.