missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%

Product Code. 10192900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192900 1 g Glass bottle 1g
10014013 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192900 Supplier Thermo Scientific Chemicals Supplier No. 111970010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 109.0°C to 111.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 57.00
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-D-galactose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.