missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2-fluoro-D-galactose, TRC

Product Code. 30179271
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179271 10 mg 10mg
30179423 50 mg 50mg
30179402 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179271 Supplier Toronto Research Chemicals Supplier No. TRCD23300010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-53-3
Molecular Formula C6 H11 F O5
Molecular Weight (g/mol) 182.15
Synonym D-Galactose, 2-deoxy-2-fluoro-, 2-Deoxy-2-fluoro-D-galactose, 2-Deoxy-2-fluoro-D-galactopyranose
IUPAC Name (3R,4S,5R,6R)-3-fluoro-6-(hydroxymethyl)oxane-2,4,5-triol
SMILES OC[C@H]1OC(O)[C@H](F)[C@@H](O)[C@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H11FO5/c7-3-5(10)4(9)2(1-8)12-6(3)11/h2-6,8-11H,1H2/t2-,3-,4+,5-,6?/m1/s1
Formula Weight 182.0591
Chemical Name or Material 2-Deoxy-2-fluoro-D-galactose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.