missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-Alpha-D-arabinofuranose, TRC

Product Code. 30179625
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179625 25 g 25g
30178921 5 g 5g
30178922 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179625 Supplier Toronto Research Chemicals Supplier No. TRCD23550025G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97614-43-2
Molecular Formula C26H21FO7
Molecular Weight (g/mol) 464.44
Synonym 2-Deoxy-2-fluoro-alpha-D-arabinofuranose 1,3,5-Tribenzoate, 2'-Fluoro-2'-deoxy-1,3,5-tri-O-benzoyl-alpha-D-arabinofuranose;
IUPAC Name [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
SMILES F[C@@H]1[C@@H](OC(=O)c2ccccc2)O[C@H](COC(=O)c3ccccc3)[C@H]1OC(=O)c4ccccc4

Specifications

Quantity 25 g
InChI Formula InChI=1S/C26H21FO7/c27-21-22(33-24(29)18-12-6-2-7-13-18)20(16-31-23(28)17-10-4-1-5-11-17)32-26(21)34-25(30)19-14-8-3-9-15-19/h1-15,20-22,26H,16H2/t20-,21+,22-,26-/m1/s1
Formula Weight 464.1271
Chemical Name or Material 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-alpha-D-arabinofuranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.