missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropylethanol, 96%

Product Code. 11397526
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397526 1 g 1g
11317536 250 mg 250mg
11307536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397526 Supplier Thermo Scientific Alfa Aesar Supplier No. L11080.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2566-44-1 | C5H10O | 86.13 g/mol

2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-44-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00040762
InChI Key LUNMJRJMSXZSLC-UHFFFAOYSA-N
Synonym cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687
PubChem CID 137642
SMILES OCCC1CC1

Specifications

Boiling Point 137°C to 138°C
Flash Point 47°C (116°F)
Refractive Index 1.4355
Quantity 1 g
UN Number UN1987
Beilstein 2036028
Solubility Information Miscible with water.
IUPAC Name 2-cyclopropylethanol
Formula Weight 86.13
Percent Purity 96%
Chemical Name or Material 2-Cyclopropylethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.