missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylphenol, TRC

Product Code. 30364652
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364652 500 mg -
30361164 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30364652 Supplier Toronto Research Chemicals Supplier No. TRCC988565500MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-84-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
Synonym o-Cyclopentylphenol, 2-Cyclopentylphenol, NSC 407789, o-Cyclopentylphenol
IUPAC Name 2-cyclopentylphenol
SMILES Oc1ccccc1C2CCCC2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H14O/c12-11-8-4-3-7-10(11)9-5-1-2-6-9/h3-4,7-9,12H,1-2,5-6H2
Formula Weight 162.1045
Chemical Name or Material 2-Cyclopentylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.