missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentyloxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 11454550
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454550 1 g 1g
11464550 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454550 Supplier Thermo Scientific Alfa Aesar Supplier No. H50110.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-90-0
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.18
MDL Number MFCD07781170
InChI Key GZCZGFSEHAZEPS-UHFFFAOYSA-N
Synonym 2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
PubChem CID 24208792
IUPAC Name 2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1

Specifications

Quantity 1 g
Formula Weight 289.19
Percent Purity 96%
Chemical Name or Material 2-Cyclopentyloxypyridine-3-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.