missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylethanol, 98+%

Product Code. 11496056
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11496056

Brand: Thermo Scientific Alfa Aesar B22344.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-00-7 | C7H14O | 114.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-00-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00019301
InChI Key JEXQWCBPEWHFKC-UHFFFAOYSA-N
Synonym cyclopentaneethanol, 2-cyclopentylethan-1-ol, 2-cyclopentaneethanol, 2-hydroxyethylcyclopentane, 2-cyclopentyl-ethanol, 2-cyclopentyl ethanol, cyclopentylethanol, cyclopentane-ethanol, 2-cyclopentanethanol, acmc-1bgun
PubChem CID 69833
IUPAC Name 2-cyclopentylethanol
SMILES C1CCC(C1)CCO

Specifications

Density 0.92
Boiling Point 182°C
Flash Point 70°C (158°F)
Refractive Index 1.4585
Quantity 1 g
Beilstein 2069860
Formula Weight 114.19
Percent Purity ≥98%
Chemical Name or Material 2-Cyclopentylethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.