missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylamine, 97%

Product Code. 11397549
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397549 1 g 1g
11307559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397549 Supplier Thermo Scientific Alfa Aesar Supplier No. H27987.03
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4442-85-7 | C8H17N | 127.23 g/mol

In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Protect from direct light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4442-85-7
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00058668
InChI Key HFACYWDPMNWMIW-UHFFFAOYSA-N
Synonym 2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride
PubChem CID 20509
IUPAC Name 2-cyclohexylethanamine
SMILES NCCC1CCCCC1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 127.23
Percent Purity 97%
Chemical Name or Material 2-Cyclohexylethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.