missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethyl-4-O-(alpha-D-glucopyranosyl)-beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Product Code. 15420597
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420597 250 mg 250mg
15410597 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420597 Supplier Thermo Scientific Alfa Aesar Supplier No. J66517.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 260804-65-7 | Molecular Formula: C20H36O11 | Molecular Weight: 452.497 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 260804-65-7
Molecular Formula C20H36O11
Molecular Weight (g/mol) 452.497
MDL Number MFCD01862984
InChI Key RHXPDNGQJSXOMW-OIIXUNCGSA-N
Synonym 2-cyclohexylethyl-4-o-a-d-glucopyranosyl-b-d-glucopyranoside, ma5, 2-6-2-cyclohexylethoxy-tetrahydro-4,5-dihydroxy-2 hydroxymethyl-2h-pyran-3-yloxy-tetrahydro-6 hydroxymethyl-2h-pyran-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-2-cyclohexylethoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cymal-2, cymal™-2, cymal r-2, 2-cyclohexylethyl beta-maltoside, 2-cyclohexylethyl, a-d-maltoside, 2-cyclohexylethyl-beta-d-maltoside
PubChem CID 9547906
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-(2-cyclohexylethoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1CCC(CC1)CCOC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)O)O)O)O)O

Specifications

Color White
Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 452.49
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Cyclohexylethyl-4-O-(alpha-D-glucopyranosyl)-beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.