missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexyl-4,6-dinitrophenol, TRC

Product Code. 30006945
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006945 100 mg 100mg
30004575 500 mg 500mg
30006783 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006945 Supplier Toronto Research Chemicals Supplier No. TRCC990029100MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-89-5
Molecular Formula C12 H14 N2 O5
Molecular Weight (g/mol) 266.25
Synonym 2-Cyclohexyl-4,6-dinitrophenol, 2,4-Dinitro-6-cyclohexylphenol, 4,6-Dinitro-o-cyclohexylphenol, DN, DN (pesticide), DN 1, DNOCHP, Dinex, Dry Mix No. 1, NSC 403662, NSC 7739, SN 46
IUPAC Name 2-cyclohexyl-4,6-dinitrophenol
SMILES Oc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C2CCCCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H14N2O5/c15-12-10(8-4-2-1-3-5-8)6-9(13(16)17)7-11(12)14(18)19/h6-8,15H,1-5H2
Formula Weight 266.0903
Chemical Name or Material 2-Cyclohexyl-4,6-dinitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.