missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-one, 97%

Product Code. 10112380 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112380 10 g Glass bottle 10g
10645912 25 g Glass bottle 25g
10395690 100 g Glass bottle 100g
11984831 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10112380 Supplier Thermo Scientific Acros Supplier No. 111240100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-68-7 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-68-7
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00001577
InChI Key FWFSEYBSWVRWGL-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one
PubChem CID 13594
ChEBI CHEBI:15977
IUPAC Name cyclohex-2-en-1-one
SMILES O=C1CCCC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -53.0°C
Color Yellow
Density 0.9930g/mL
Boiling Point 168.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H8(=O)
Refractive Index 1.4875 to 1.4895
Quantity 10 g
Beilstein 07,II,55
Fieser 01,18
Specific Gravity 0.993
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyclohexen-1-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.