missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-ol, 95%

Product Code. 10139979 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10139979 5 g Glass bottle 5g
10656260 1 g Glass bottle 1g
10413170 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10139979 Supplier Thermo Scientific Acros Supplier No. 111230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-67-3 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-67-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00001570
InChI Key PQANGXXSEABURG-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-ol, cyclohex-2-enol, 2-cyclohexene-1-ol, 2-cyclohexenol, 3-hydroxycyclohexene, 1-cyclohexen-3-ol, cyclohexen-3-ol, cyclohex-2-ene-1-ol, 1,2,3,4-tetrahydrophenol, cyclohexen-3-ol van
PubChem CID 13198
IUPAC Name cyclohex-2-en-1-ol
SMILES C1CC=CC(C1)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.9840g/mL
Boiling Point 164.0°C to 166.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H9OH
Refractive Index 1.4855
Quantity 5 g
Beilstein 06,II,6
Specific Gravity 0.984
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyclohexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.