missing translation for 'onlineSavingsMsg'
Learn More

(2-Cyanoethyl)triethoxysilane, 97%

Product Code. 11302216
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11302216

Brand: Thermo Scientific Alfa Aesar L03543.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 919-31-3 | C9H19NO3Si | 217.34 g/mol

(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-31-3
Molecular Formula C9H19NO3Si
Molecular Weight (g/mol) 217.34
MDL Number MFCD00019851
InChI Key GBQYMXVQHATSCC-UHFFFAOYSA-N
Synonym 3-triethoxysilyl propionitrile, 2-cyanoethyl triethoxysilane, 2-cyanoethyltriethoxysilane, propanenitrile, 3-triethoxysilyl, beta-cyanoethyltriethoxysilane, 3-triethoxysilyl propanenitrile, propionitrile, 3-triethoxysilyl, silane, 2-cyanoethyl triethoxy, triethoxy-2-kyanethylsilan, unii-0i8r6d195f
PubChem CID 13522
IUPAC Name 3-triethoxysilylpropanenitrile
SMILES CCO[Si](CCC#N)(OCC)OCC

Specifications

Boiling Point 224°C to 225°C
Quantity 25 g
Beilstein 1776392
Flash Point 100°C (212°F)
Solubility Information Hydrolyzes in water.
Formula Weight 217.34
Percent Purity 97%
Refractive Index 1.413
Sensitivity Moisture sensitive
Density 0.98
Chemical Name or Material (2-Cyanoethyl)triethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.