missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite, 95%

Product Code. 15198677 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15198677 5 g 5g
15234714 1 g 1g
15244714 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15198677 Supplier Thermo Scientific Acros Supplier No. 379740050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102691-36-1 | C15H32N3OP | 301.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102691-36-1
Molecular Formula C15H32N3OP
Molecular Weight (g/mol) 301.41
InChI Key RKVHNYJPIXOHRW-UHFFFAOYSA-N
Synonym 3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile
PubChem CID 128153
IUPAC Name 3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
SMILES CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N

Specifications

Density 0.9490g/mL
Boiling Point 98.0°C to 100.0°C (5.0 mmHg)
Flash Point 60°C
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.47
Quantity 5 g
Specific Gravity 0.949
Formula Weight 301.41
Percent Purity 95%
Chemical Name or Material 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.