missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyl N,N,N',N'-Tetraisopropylphosphoramidite, TRC

Product Code. 30359703
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359703 5 g 5g
30365548 1 g 1g
30360444 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359703 Supplier Toronto Research Chemicals Supplier No. TRCC9821005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102691-36-1
Molecular Formula C15 H32 N3 O P
Molecular Weight (g/mol) 301.41
IUPAC Name 3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
SMILES CC(C)N(C(C)C)P(OCCC#N)N(C(C)C)C(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H32N3OP/c1-12(2)17(13(3)4)20(19-11-9-10-16)18(14(5)6)15(7)8/h12-15H,9,11H2,1-8H3
Formula Weight 301.2283
Chemical Name or Material 2-Cyanoethyl N,N,N',N'-Tetraisopropylphosphoramidite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.