missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzoic Acid, TRC

Product Code. 30011133
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011133 500 mg 500mg
30006266 5 g 5g
30008043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011133 Supplier Toronto Research Chemicals Supplier No. TRCC962353500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3839-22-3
Molecular Formula C8 H5 N O2
Molecular Weight (g/mol) 147.131
Synonym 2-Carboxybenzonitrile, NSC 193443, o-Carboxybenzonitrile, o-Cyanobenzoic Acid;
IUPAC Name 2-cyanobenzoic acid
SMILES OC(=O)c1ccccc1C#N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H5NO2/c9-5-6-3-1-2-4-7(6)8(10)11/h1-4H,(H,10,11)
Formula Weight 147.032
Chemical Name or Material 2-Cyanobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.