missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzeneboronic acid, 96%

Product Code. 15459367
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459367 1 g 1g
15469367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459367 Supplier Thermo Scientific Alfa Aesar Supplier No. H52609.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138642-62-3 | C7H6BNO2 | 146.94 g/mol

2-Cyanobenzeneboronic acid is useful in the synthesis of substitutes indenones or indanones. It is used in a rhodium-catalyzed [3+2] annulation with alkynes leading to substituted indenones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzeneboronic acid is useful in the synthesis of substitutes indenones or indanones. It is used in a rhodium-catalyzed [3+2] annulation with alkynes leading to substituted indenones.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138642-62-3
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01632208
InChI Key NPLZNDDFVCGRAG-UHFFFAOYSA-N
Synonym 2-cyanophenyl boronic acid, 2-cyanobenzeneboronic acid, 2-cyanophenylboric acid, 2-cyanophenylboronicacid, 2-dihydroxyboranyl benzonitrile, boronic acid, b-2-cyanophenyl, 2-boronobenzonitrile, 2-cyanobenzene boronic acid, 2-cyano-phenyl-boronic acid, boronic acid, cyanophenyl
PubChem CID 2734610
IUPAC Name (2-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C#N

Specifications

Melting Point 250°C to 254°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 146.94
Percent Purity 96%
Chemical Name or Material 2-Cyanobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.