missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzene-1-sulfonyl chloride, ≥95%, Maybridge

Product Code. 10400964
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10400964 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10400964 Supplier Thermo Scientific Maybridge Supplier No. TL00431EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 69360-26-5
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C7H4ClNO2S
MDL Number MFCD00052341
Quantity 25 g
Synonym 2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7
InChI Key NQAYCMBZPAARNO-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl
IUPAC Name 2-cyanobenzenesulfonyl chloride
Molecular Weight (g/mol) 201.624
PubChem CID 522347
Formula Weight 201.63
Percent Purity 95%
Chemical Name or Material 2-Cyanobenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.