missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, TRC

Product Code. 30133725
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133725 10 g 10g
30135721 50 g 50g
30011904 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133725 Supplier Toronto Research Chemicals Supplier No. TRCC98224510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24522-30-3
Molecular Formula C10 H7 F3 N2 O
Molecular Weight (g/mol) 228.17
Synonym 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, Leflunomide Imp. H (EP)
IUPAC Name 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
SMILES FC(F)(F)c1ccc(NC(=O)CC#N)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H7F3N2O/c11-10(12,13)7-1-3-8(4-2-7)15-9(16)5-6-14/h1-4H,5H2,(H,15,16)
Formula Weight 228.051
Chemical Name or Material 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.