missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methylpyridine, 97%

Product Code. 10192914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10192914 5 g 5g
10173154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192914 Supplier Thermo Scientific Acros Supplier No. 425920050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1620-75-3 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1620-75-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00190586
InChI Key CMADFEQMYFNYCF-UHFFFAOYSA-N
Synonym 6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril
PubChem CID 74182
IUPAC Name 6-methylpyridine-2-carbonitrile
SMILES CC1=CC=CC(=N1)C#N

Specifications

Melting Point 70.0°C to 74.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 2-Cyano-6-methylpyridine
compliance-icons
Product Identifier
  • 2-Cyano-6-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.