missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methoxybenzothiazole, 99%, Thermo Scientific™

Product Code. 10167883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
250mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10167883 250mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10167883 Supplier Thermo Scientific Acros Supplier No. 225362500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 943-03-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 131.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H6N2OS
MDL Number MFCD00010537
Quantity 250 mg
Synonym 2-cyano-6-methoxybenzothiazole, 6-methoxybenzo d thiazole-2-carbonitrile, 6-methoxy-2-benzothiazolecarbonitrile, 6-methoxybenzothiazole-2-carbonitrile, 6-methoxy-benzothiazole-2-carbonitrile, 6-methoxy-2-cyano-benzothiazole, 2-benzothiazolecarbonitrile, 6-methoxy, pubchem9766, acmc-209rr8, ksc486m5p
InChI Key DEWDWBYQOFXKIH-UHFFFAOYSA-N
SMILES COC1=CC2=C(C=C1)N=C(S2)C#N
IUPAC Name 6-methoxy-1,3-benzothiazole-2-carbonitrile
Molecular Weight (g/mol) 190.23
PubChem CID 342109
Formula Weight 190.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyano-6-methoxybenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.