missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 15441719
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95727-86-9
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD01001119
InChI Key WDSCJULUXJSJOX-UHFFFAOYSA-N
Synonym 5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
PubChem CID 1535372
IUPAC Name 5-(trifluoromethyl)pyridine-2-carbonitrile
SMILES FC(F)(F)C1=CN=C(C=C1)C#N

Specifications

Quantity 1 g
UN Number UN3439
Beilstein 13339826
Solubility Information Slightly soluble in water.
Formula Weight 172.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Cyano-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'