missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10202842
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202842 1 g Amber Glass Bottle 1g
10482213 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10202842 Supplier Thermo Scientific Maybridge Supplier No. MO00753DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 197960-31-9
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.65
MDL Number MFCD03659720
InChI Key ZFQDREVAYVNITB-UHFFFAOYSA-N
Synonym benzenesulfonylchloride, 2-cyano-5-methyl, 2-cyano-5-methyl-benzenesulfonyl chloride, 2-cyano-5-methylbenzene-1-sulfonyl chloride, 3-chlorosulphonyl-4-cyanotoluene, 2-cyano-5-methylbenzenesulfonylchloride, 2-chlorosulphonyl-4-methylbenzonitrile, 2-cyano-5-methylbenzene-sulfonyl chloride, 2-cyano-5-methylbenzenesulphonyl chloride, 2-cyano-5-methylphenylsulfonyl chloride, 2-chlorosulfonyl-4-methylbenzenecarbonitrile
PubChem CID 2779837
SMILES CC1=CC(=C(C=C1)C#N)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-cyano-5-methylbenzene-1-sulfonyl chloride
Formula Weight 215.66
Percent Purity 97%
Chemical Name or Material 2-Cyano-5-methylbenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.