missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4-methylpyridine, 98%

Product Code. 11353976
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353976 5 g 5g
11343976 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353976 Supplier Thermo Scientific Alfa Aesar Supplier No. L17148.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1620-76-4 | C7H6N2 | 118.139 g/mol

2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1620-76-4
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00128868
InChI Key LQAWSWUFSHYCHP-UHFFFAOYSA-N
Synonym 2-cyano-4-methylpyridine, 4-methylpicolinonitrile, 4-methyl-2-pyridinecarbonitrile, 2-cyano-4-picoline, 2-pyridinecarbonitrile, 4-methyl, pubchem21387, acmc-209dnq, 2-cyano-4methylpyridine, 2-cyano 4-methylpyridine, 4-methyl-2-cyanopyridine
PubChem CID 88739
IUPAC Name 4-methylpyridine-2-carbonitrile
SMILES CC1=CC(=NC=C1)C#N

Specifications

Melting Point 85°C to 88°C
Boiling Point 145°C to 148°C (38 mmHg)
Odor Characteristic
Quantity 5 g
Beilstein 110753
Solubility Information Slightly soluble in water.
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 2-Cyano-4-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.